Order Entry
Germany
ContactUsLinkComponent
 
Diethyl ether, dried (max. 0.0075% H₂O) stabilised, Puriss. p.a. ACS, Riedel-de Haën™
Catalog # HONC34942-18L
CAS Number:  
Diethyl ether, dried (max. 0.0075% H₂O) stabilised, Puriss. p.a. ACS, Riedel-de Haën™
Catalog # HONC34942-18L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can, returnable
  • Pk:
    18 L
  • Seq:
    5267

Specifications

Specification Test Results

  • ExplosionLimit:
    36.5 %
  • Assay (GC):
    Min. 99.5 %
  • Boiling range:
    34 - 35 °C
  • Density (D 20/4):
    0.713 - 0.714
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.0075 %
  • Free acid (as CH₃COOH):
    Max. 0.0002 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.000005 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.000005 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.000005 %
  • Acetone (GC):
    Max. 0.005 %
  • Aldehydes:
    complying
  • Carbonyl compounds (as CO):
    Max. 0.001 %
  • Ethanol (GC):
    Max. 0.02 %
  • Methanol (GC):
    Max. 0.02 %
  • Peroxides (as H₂O₂):
    Max. 0.00003 %
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10

Specification Test Results

About this item

 :
 :74.12 g/mol
Boiling Pt:34…35 °C (1,013 hPa)
Melting Pt:–116 °C.(1,013 hPa)
Density:0.71 g/cm3 (20 °C)
Flash Pt:–45 °C (1,013 hPa.)
Storage Temperature:Ambient
UN:1155
ADR:3,I
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646