Order Entry
Germany
Orders LinkContactUsLinkComponent
Zolmitriptan Related Compounds, Reference Standard

Zolmitriptan Related Compounds, Reference Standard

Supplier:   USP
Zolmitriptan Related Compounds, Reference Standard
Supplier:  USP
Certificates
Zolmitriptan Related Compounds, Reference Standard

6 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1727020.
  • 1727031.
  • 1727064.
  • 1727075.
  • 1727086.
  • 1727097.
  • Description:
  • Zolmitriptan Related Compound A
  • Zolmitriptan Related Compound B
  • Zolmitriptan Related Compound E
  • Zolmitriptan Related Compound F
  • Zolmitriptan Related Compound G
  • Zolmitriptan Related Compound H
  • Synonym(s):
  • (S)-4-({3-[2-(Methylamino)ethyl]-1H-indol-5-yl}methyl)oxazolidin-2-one
  • (S)-2-Amino-3-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}propan-1-ol
  • (S)-N,N-dimethyl-2-{5-[(2-oxooxazolidin-4-yl)methyl]-1H-indol-3-yl}ethanamine oxide
  • 2,2'-[4-(Dimethylamino)butane-1,1-diyl]bis{5-[(S)-(2-oxooxazolidin-4-yl)methyl]-3-(2-dimethylaminoethyl)indole}
  • (S)-4-(4-Aminobenzyl)oxazolidin-2-one
  • 4,4-Diethoxy-N,N-dimethylbutan-1-amine
  • CAS No.:
  • 139264-35-0
  • 139264-69-0
  • 251451-30-6
  • 1350928-05-0
  • 152305-23-2
  • 1116-77-4
  • UN No.:
  • 3267

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1727020.
    Zolmitriptan Related Compound A
    Zolmitriptan Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 2 540,00 €
    2 540,00 €
    1 * 20 mg
       2 540,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727020.
    Description
    Zolmitriptan Related Compound A
    Synonym(s)
    (S)-4-({3-[2-(Methylamino)ethyl]-1H-indol-5-yl}methyl)oxazolidin-2-one
    CAS No.
    139264-35-0
    UN No.
  • 1727031.
    Zolmitriptan Related Compound B
    Zolmitriptan Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 220,00 €
    1 220,00 €
    1 * 20 mg
       1 220,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727031.
    Description
    Zolmitriptan Related Compound B
    Synonym(s)
    (S)-2-Amino-3-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}propan-1-ol
    CAS No.
    139264-69-0
    UN No.
  • 1727064.
    Zolmitriptan Related Compound E
    Zolmitriptan Related Compound E
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 140,00 €
    1 140,00 €
    1 * 25 mg
       1 140,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727064.
    Description
    Zolmitriptan Related Compound E
    Synonym(s)
    (S)-N,N-dimethyl-2-{5-[(2-oxooxazolidin-4-yl)methyl]-1H-indol-3-yl}ethanamine oxide
    CAS No.
    251451-30-6
    UN No.
  • 1727075.
    Zolmitriptan Related Compound F
    Zolmitriptan Related Compound F
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 250,00 €
    1 250,00 €
    1 * 25 mg
       1 250,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727075.
    Description
    Zolmitriptan Related Compound F
    Synonym(s)
    2,2'-[4-(Dimethylamino)butane-1,1-diyl]bis{5-[(S)-(2-oxooxazolidin-4-yl)methyl]-3-(2-dimethylaminoethyl)indole}
    CAS No.
    1350928-05-0
    UN No.
  • 1727086.
    Zolmitriptan Related Compound G
    Zolmitriptan Related Compound G
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 320,00 €
    1 320,00 €
    1 * 25 mg
       1 320,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727086.
    Description
    Zolmitriptan Related Compound G
    Synonym(s)
    (S)-4-(4-Aminobenzyl)oxazolidin-2-one
    CAS No.
    152305-23-2
    UN No.
  • 1727097.
    Zolmitriptan Related Compound H
    Zolmitriptan Related Compound H
    USP reference standard
    Ampule
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 1 330,00 €
    1 330,00 €
    1 * 50 mg
       1 330,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1727097.
    Description
    Zolmitriptan Related Compound H
    Synonym(s)
    4,4-Diethoxy-N,N-dimethylbutan-1-amine
    CAS No.
    1116-77-4
    UN No.
    3267
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.