Order Entry
Germany
Orders LinkContactUsLinkComponent
Clarithromycin Related Compounds, Reference Standard

Clarithromycin Related Compounds, Reference Standard

Clarithromycin Related Compound A, USP reference standard, Vial, 25 mg
Catalog # 1134380.
Supplier:   USP
Clarithromycin Related Compounds, Reference Standard
Catalog # 1134380.
Supplier:  USP
CAS Number:  
Clarithromycin Related Compounds, Reference Standard

Clarithromycin Related Compound A, USP reference standard, Vial, 25 mg

CAS Number:  
Price
1 140,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    USP reference standard
  • Description:
    Clarithromycin Related Compound A
  • Cat. No.:
    1134380.
  • Formula:
    C₃₉H₇₁NO₁₃
  • Description:
    Clarithromycin Related Compound A
  • Synonym(s):
    6,11-di-O-methylerythromycin A
  • CAS No.:
    81103-14-2

Specifications