Rivaroxaban Phthalamide, Pharmaceutical Analytical Impurities
1A00660.
: USP
:
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1365267-36-2
- Description:Rivaroxaban Phthalamide
- Synonym(s):N1,N2-bis(((S)-2-oxo-3-(4-(3-oxomorpholino)phenyl)oxazolidin-5-yl)methyl)phthalamide
- Description:Rivaroxaban Phthalamide
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.