Order Entry
Germany
Orders LinkContactUsLinkComponent
Methimazole, Reference Standards

Methimazole, Reference Standards

Supplier:   USP
Methimazole, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1411005.
  • 1411016.
  • 1411038.
  • Description:
  • Methimazole
  • Methimazole Related Compound A
  • Methimazole Related Compound C
  • Synonym(s):
  • 2,2-Dimethoxy-N-methylethanamine
  • 1-Methyl-2-(methylthio)-1H-imidazole
  • CAS No.:
  • 60-56-0
  • 122-07-6
  • 14486-52-3
  • UN No.:
  • 1989

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1411005.
    Methimazole
    Methimazole
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 404,00 €
    404,00 €
    1 * 200 mg
       404,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1411005.
    Description
    Methimazole
    Synonym(s)
    CAS No.
    60-56-0
    UN No.
  • 1411016.
    Methimazole Related Compound A
    Methimazole Related Compound A
    USP reference standard
    3 x 50 mg
    Ampule
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 1 250,00 €
    1 250,00 €
    1 * 150 mg
       1 250,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1411016.
    Description
    Methimazole Related Compound A
    Synonym(s)
    2,2-Dimethoxy-N-methylethanamine
    CAS No.
    122-07-6
    UN No.
    1989
  • 1411038.
    Methimazole Related Compound C
    Methimazole Related Compound C
    USP reference standard
    3 x 50 mg
    Ampule
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - 1 260,00 €
    1 260,00 €
    1 * 150 mg
       1 260,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1411038.
    Description
    Methimazole Related Compound C
    Synonym(s)
    1-Methyl-2-(methylthio)-1H-imidazole
    CAS No.
    14486-52-3
    UN No.