Order Entry
Germany
Orders LinkContactUsLinkComponent
Ibutilide Related Compounds, Reference Standard

Ibutilide Related Compounds, Reference Standard

Supplier:   USP
Ibutilide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Ibutilide Related Compounds, Reference Standard

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1335610.
  • 1335621.
  • 1335632.
  • Description:
  • Ibutilide Fumarate
  • Ibutilide Related Compound A
  • Ibutilide Related Compound B
  • Synonym(s):
  • N-Ethyl-N-heptyl-4-[4-(methylsulfonamido)phenyl]-4-oxobutanamide
  • N-Ethyl-N-heptyl-4-hydroxy-4-[4-(methylsulfonamido)phenyl]butanamide
  • CAS No.:
  • 122647-32-9
  • 100632-58-4
  • 160087-98-9

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1335610.
    Ibutilide Fumarate
    Ibutilide Fumarate
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 959,00 €
    959,00 €
    1 * 100 mg
       959,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1335610.
    Description
    Ibutilide Fumarate
    Synonym(s)
    CAS No.
    122647-32-9
  • 1335621.
    Ibutilide Related Compound A
    Ibutilide Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 507,00 €
    507,00 €
    1 * 20 mg
       507,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1335621.
    Description
    Ibutilide Related Compound A
    Synonym(s)
    N-Ethyl-N-heptyl-4-[4-(methylsulfonamido)phenyl]-4-oxobutanamide
    CAS No.
    100632-58-4
  • 1335632.
    Ibutilide Related Compound B
    Ibutilide Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 507,00 €
    507,00 €
    1 * 20 mg
       507,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1335632.
    Description
    Ibutilide Related Compound B
    Synonym(s)
    N-Ethyl-N-heptyl-4-hydroxy-4-[4-(methylsulfonamido)phenyl]butanamide
    CAS No.
    160087-98-9