
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Catalog # BC23
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Spacer Arm (Å):11.6
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:No
- Water Soluble:Yes
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:436.37 g/mol
:GHS07
CAS Number:92921-24-9
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978