Order Entry
Germany
ContactUsLinkComponent
Sulpho-NHS-LC-Biotin, EZ-Link™ No-Weigh™ Format, Pierce™
Sulpho-NHS-LC-Biotin, EZ-Link™ No-Weigh™ Format, Pierce™
Catalog # PIERA39257
CAS Number:  
Sulpho-NHS-LC-Biotin, EZ-Link™ No-Weigh™ Format, Pierce™
Catalog # PIERA39257
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275
  • Pk Info:
    10×1 mg

Specifications

Specification Test Results

  • Product Type:
    Sulfo-NHS-LC-Biotin
  • Quantity:
    10 x 1 mg
  • Labeling Method:
    Chemical Labeling
  • Shipping Condition:
    Qualified for Ambient or Wet Ice
  • Product Line:
    EZ-Link™
  • Cell Permeability:
    Cell-Impermeant
  • Chemical Reactivity:
    Amine
  • Reactive Moiety:
    NHS Ester
  • Solubility:
    DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
  • Label or Dye:
    Biotin
  • Spacer:
    Mid-Length

Specification Test Results

About this item

 :
 :556.59 g/mol
Storage Temperature:Freezer
CAS Number:127062-22-0
Formula:C₂₀H₂₉N₄NaO₉S₂
MDL Number:MFCD00078534