Order Entry
Germany
ContactUsLinkComponent
Sulfo-NHS-LC-LC-Biotin, EZ-Link™, Pierce™
Sulfo-NHS-LC-LC-Biotin, EZ-Link™, Pierce™
Certificates
undefined
Sulfo-NHS-LC-LC-Biotin, EZ-Link™, Pierce™

Specification Test Results

  • Product Type:
    Sulfo-NHS-LC-LC-Biotin
  • Quantity:
    50 mg
  • Product Line:
    EZ-Link™
  • Cell Permeability:
    Cell-Impermeant
  • Chemical Reactivity:
    Amine
  • Reactive Moiety:
    Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
  • Solubility:
    DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
  • Label or Dye:
    Biotin
  • Label Type:
    Biotin & Analogs
  • Spacer:
    Long
  • Spacer arm:
    22.4 angstroms

Specification Test Results