SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate), Pierce™
Catalog # PIER22416
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Maleimide, NHS Ester
- Spacer:Mid-Length
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMSO
Specification Test Results
About this item
:
:356.33 g/mol
Storage Temperature:Freezer
CAS Number:79886-55-8
Formula:C₁₈H₁₆N₂O₆
MDL Number:MFCD00054961