Order Entry
Germany
ContactUsLinkComponent
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥97% (by HPLC), Millipore®
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥97% (by HPLC), Millipore®
Catalog # 573114-100
CAS Number:  
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate) ≥97% (by HPLC), Millipore®
Catalog # 573114-100
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :334.33 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Freezer
CAS Number:64987-85-5
Formula:C₁₆H₁₈N₂O₆
MDL Number:MFCD00009634