Orders LinkContactUsLinkComponent
Micro-spatula samplers, SteriWare
Micro-spatula samplers, SteriWare
Micro-spatula samplers, SteriWare
Supplier:  Sampling Systems
Certificates
Micro-spatula samplers, SteriWare
Micro-spatula samplers, SteriWare
4 Options Available Below
Product Details & Documents
Disposable Micro-Spatulas are ideal for use with small amounts of powder and granule.
  • Ideal for sampling, dispensing, and measuring
  • Excellent for use in confined spaces
  • Available in two colours and multiple sizes
  • Easy to use
Sampling small amounts of powder or granule is made easy with the SteriWare Micro-Spatula. This single-use spatula is excellent when needing to take samples from confined spaces, and it eliminates the need for costly cleaning and cleaning validation.
Product Family Options
Product Information
  • Material
    Colour
    Volume
    Availability
    Price
  • 300-0489
    Sterile micro-spatula, 4 mm blade width
    Sterile micro-spatula, 4 mm blade width
    PP
    Blue
    0,3 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 225,00 €
    225,00 €
    1 * 100 ST
       225,00 €
  • 300-0490
    Sterile micro-spatula, 6 mm blade width
    Sterile micro-spatula, 6 mm blade width
    PP
    Blue
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 181,00 €
    181,00 €
    1 * 100 ST
       181,00 €
  • 300-0491
    Sterile micro-spatula, 13 mm blade width
    Sterile micro-spatula, 13 mm blade width
    PP
    Blue
    2 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 225,00 €
    225,00 €
    1 * 100 ST
       225,00 €
  • 300-0488
    Sterile micro-spatula, 13 mm blade width
    Sterile micro-spatula, 13 mm blade width
    PS
    Red
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 189,00 €
    189,00 €
    1 * 1 ST
       189,00 €