Order Entry
Germany
ContactUsLinkComponent
Epoxomicin (synthetic) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Epoxomicin (synthetic) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 324800-100
CAS Number:  
Epoxomicin (synthetic) ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 324800-100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
 :554.7 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:134381-21-8
Formula:C₂₈H₅₀N₄O₇
MDL Number:MFCD03791061