ContactUsLinkComponent
Earplugs, Tri-Flange™
Earplugs, Tri-Flange™
Catalog # MMMAPN01006
Supplier:  3M
undefined
Earplugs, Tri-Flange™
Catalog # MMMAPN01006
Supplier:  3M
Specifications
  • Description:
    E-A-R™ Tri-Flange™ earplugs, 29 dB, cloth cord
  • Pk:
    400 PAIR
Specifications
About this item
3M™ Tri-Flange™ Earplugs are innovative, comfortable hearing protectors with soft, flexible flanges that adapt to the shape of ears.
  • Reduces noise levels up to 29 dB
  • Features soft, flexible triple flanges that adapt to the shape of the ear for an effective acoustic seal
  • No need to handle the plug tip, simply push the plug to fit
  • Ideal for use in dirty environments
  • Easy to wash and reuse
  • Cloth cord
Get comfortable and hygienic hearing protection with 3M™ Tri-Flange™ Earplugs. Reliable and robust, the plugs are moisture-resistant making them suitable for use multiple times and easily washed. CE Certified, they are approved against EN 352-2 and reduce noise levels up to 29 dB.
Included cloth cord keeps the plugs together and can be threaded around the neck, convenient when moving in and out of noisy environments.