Safety goggles 4800 series
Catalog # MMMABUD48AF
Supplier: 3M
Specifications
- Type:Indirect venting, anti-fog
- Description:Safety goggles, 4800 series
- Lenses:Clear PC
- Pk:1
Specifications
About this item
3M™ 4800 series safety goggles feature a clear, strong polycarbonate lens for impact resistance and indirect ventilation for added comfort.
- Basic economical safety goggle with a flat lens design
- Indirect ventilation to protect against liquid droplets
- Polycarbonate lens for impact resistance
- Not designed for wear over prescription glassess
- Anti-fog coating to help reduce lens fogging
For all-round safety goggles which provide cooling comfort, the 4800 Series features an indirect ventilation system to prevent misting and ensures comfort while working over long periods. They also have a clear, strong polycarbonate lens to protect against impacts and flying objects. They are available with an anti-fog lens coating.