SARS-CoV-2 (2019-nCoV) Nucleocapsid-His Recombinant Protein
Catalog # SINO40588-V07E-100
Supplier: Sino Biological
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:SARS-CoV-2
- Storage conditions:–20…–80 °C
- Endotoxin content:Please contact us for more information.
- Gene ID:YP_009724397.2
- Reconstitution instructions:A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
- Carrier-free:Y
- Animal-free:Y
- Protein synonyms:NP-CoV
- Protein/peptide name:NP-CoV
- Purity:> 90 % as determined by SDS-PAGE.
- Molecular weight:The recombinant SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (His tag) consists of 426 amino acids and predicts a molecular mass of 46.61 kDa.
- Sequence:Met1-Ala419(335Gly/Ala)
- Formulation:Powder
- Shipping temperature:Ambient
Specifications
About this item
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Nucleocapsid Protein (YP_009724397.2) (Met1-Ala419(335Gly/Ala)) was expressed with a polyhistidine tag at the N-terminus.