Anti-IL-22BP Mouse monoclonal antibody [clone: 9A11E4F7]
Supplier: Sino Biological
Certificates
About this item
Anti-IL-22BP Antibody
Type: Primary
Antigen: IL-22BP
Clonality: Monoclonal
Clone: Clone: 9A11E4F7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Specifications
- Catalog No:
- SINO11025-MM05-200
- SINO11025-MM05-50
- Antigen symbol:
- IL-22BP
- IL-22BP
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- Clone: 9A11E4F7
- Clone: 9A11E4F7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Form:
- liquid
- liquid
- Storage temperature:
- Can be stored at 2...8 ℃. Stable for twelve months at -20...-80 ℃. Sodium azide is recommended to avoid contamination (final concentration 0.05...0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Can be stored at 2...8 ℃. Stable for twelve months at -20...-80 ℃. Sodium azide is recommended to avoid contamination (final concentration 0.05...0.1%). It is toxic to cells and should be disposed of properly. Avoid repeated freeze-thaw cycles.
- Shipping temperature:
- ambient temperature
- ambient temperature