Order Entry
Germany
ContactUsLinkComponent
Anti-GPR30 rabbit polyclonal antibody
Anti-GPR30 rabbit polyclonal antibody
  NOVUNBP1-31239
Anti-GPR30 rabbit polyclonal antibody
  NOVUNBP1-31239

 

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-coupled Receptor 30
  • Antigen symbol:
    GPER/GPR30
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human
  • Western blot:
    Yes
  • Gene ID:
    2852
  • Antigen synonyms:
    heptahelix receptor|CMKRL2MGC99678|leucine rich protein in GPR30 3'UTR|G protein-coupled estrogen receptor 1|G protein-coupled receptor 30|GPR30GPCR-Br|chemokine receptor-like 2|LERGU|Flow-induced endothelial G-protein coupled receptor 1|G-protein coupled estrogen receptor 1|constitutively expressed peptide-like receptor|Chemoattractant receptor-like 2|LyGPR|LERGU2|DRY12IL8-related receptor DRY12|CEPRG-protein coupled receptor 30|FEG-1mER|Lymphocyte-derived G-protein coupled receptor|Membrane estrogen receptor|GPCR-BR
  • Storage buffer:
    PBS (pH 7,0) and 20% Glycerol
  • Storage temperature:
    Aliquot and store at −20 or −80 °C. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GPR30. The exact sequence is proprietary.
  • Purification:
    Immunogen affinity purified
  • Pk:
    100 µl

 

 

Rabbit Polyclonal GPR30 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:500

Type: Primary
Antigen: GPR30
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human