Order Entry
Germany
ContactUsLinkComponent
Human recombinant enolase 1 (from E. coli)
Human recombinant enolase 1 (from E. coli)
Catalog # NOVUNBP1-30262
Supplier:  Novus Biologicals
CAS Number:  
Human recombinant enolase 1 (from E. coli)
Catalog # NOVUNBP1-30262
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    0,1 mg
  • Conjugation:
    Unconjugated
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
  • Endotoxin content:
    <1,0 EU per 1 microgram of protein (determined by LAL method)
  • Enzyme name:
    Enolase 1
  • Enzyme synonyms:
    2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin
  • Purity:
    >90%, by SDS-PAGE
  • Concentration:
    1 mg/ml
  • Tested applications:
    ELISA

Specifications

About this item

The Recombinant Human Enolase 1 Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Enolase 1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

  • Entrez Gene ID: 2023

Tested Applications: ELISA, SDS-Page. Tested Reactivity: Human.