Mouse monoclonal anti-NKG2D antibody (1D11)
Catalog # NOVUNB100-65956
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Killer Cell Lectin Like Receptor K1
- Antigen symbol:NKG2D/CD314
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Gene ID:22914
- Antigen synonyms:NK cell receptor D|NKG2-D type II integral membrane protein|CD314|KLR|NKG2-D|Killer cell lectin-like receptor subfamily K member 1|NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence|member 1|FLJ17759|killer cell lectin-like receptor subfamily K|FLJ75772|CD314 antigen|D12S2489E|NKG2-D-activating NK receptor
- Storage buffer:Phosphate buffered saline
- Storage temperature:Store at −20 °C. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:NKL cells.
- Purification:Protein G purified
- Pk:0,25 mg
Specifications
About this item
The NKG2D / CD314 Antibody (1D11) from Novus Biologicals is a mouse monoclonal antibody to NKG2D / CD314. This antibody reacts with human. The NKG2D / CD314 Antibody (1D11) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: NKG2D/CD314
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human