Order Entry
Germany
ContactUsLinkComponent
Sodium standard solutions (NIST)
Sodium standard solutions (NIST)
Catalog # HACH2318153
Supplier:  Hach
CAS Number:  
undefined
Sodium standard solutions (NIST)
Catalog # HACH2318153
Supplier:  Hach
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pk:
    1 L
  • Element:
    Sodium

Specifications

About this item

For accuracy checks and instrument calibrations in sodium determinations.

Caution: 100 mg/l = 100 µg/ml