Order Entry
Germany
ContactUsLinkComponent
Fetal bovine serum, heat inactivated, certified, U.S. origin
Fetal bovine serum, heat inactivated, certified, U.S. origin
Catalog # ROCKFBS-01-0100
Fetal bovine serum, heat inactivated, certified, U.S. origin
Catalog # ROCKFBS-01-0100

Specifications

  • Sterility:
    Sterile
  • Description:
    Fetal bovine serum, heat inactivated, certified, U.S. origin
  • Endotoxin:
    <1,0 EU/ml
  • Pk:
    100 mL

Specifications

About this item

Fatal bovine serum is used in a wide range of cell culture applications.

  • Sterile bacteriophage
  • Store at −20 °C

This bovine serum is certified to originate in the United States as heat inactivated, performance, mycoplasma, virus, bacteriophage and endotoxin tested.

Caution: For in vitro diagnostic use only, not for human or animal therapeutic use.