ContactUsLinkComponent
Powered air respirator back pack adapters, Versaflo™
Powered air respirator back pack adapters, Versaflo™
Catalog # MMMATR-655
Supplier:  3M
undefined
Powered air respirator back pack adapters, Versaflo™
Catalog # MMMATR-655
Supplier:  3M
Specifications
  • Description:
    Back pack adapter
  • Pk:
    1
Specifications
About this item
The Versaflo™ Backpack Adaptor TR-655 allows users to wear Versaflo™ Powered Air Turbo TR-600 or TR-800 unit on their back when used with Versaflo™ Backpack Harness BPK-01.
  • Effective alternative to wearing the powered air turbo on a belt
  • Designed for use alongside Versaflo™ Backpack Harness BPK-01
This adaptor is specially designed for use with Versaflo™ Powered Air Turbo and Versaflo™ Backpack Harness BPK-01. It offers users the versatility to choose how to carry their powered air turbo according to the task they’re completing.