Slip-resistant conformable tape, 500 series, Safety-Walk™
Supplier: 3M
Certificates
About this item
3M™ Safety-Walk™ Slip-Resistant Conformable Tape 500 Series is a high friction, slip-resistant material that has a special aluminium backing to conform around edges and adhere to irregular surfaces.
- Anti-slip tape for irregular or contoured surfaces in light to heavy shoe-traffic areas
- Suitable for application in dry, wet, or oily environments
- Aggressive, high performance adhesive forms an excellent bond for a secure hold
- Tapes and treads resist wrinkling, curling, tearing, shrinking, and lifting
Primarily for use on dimpled or patterned surfaces in industrial, food service, or commercial areas. The aluminum backing conforms to corners, curves, and irregular dimpled or patterned surfaces like diamond plating, riveted plates, and ladder rungs. Use the safety yellow colour for high visibility and easy identification of hazard areas.