Fragrance allergen mix A1, 24 components in tert-Butyl methyl ether, certified reference material, TraceCERT®, Supelco®
Supplier: Merck
Certificates
About this item
Ready to use certified reference material mixture suitable as calibrant for te testing of allergenic fragrances and their isomers according th the IFRA method. 24 analytes in tert-Butyl methyl ether (MTBE)
- Certification of each individual component by qNMR (following ISO/IEC 17025 accreditation)
- Mixes produced following an established workflow according to ISO 17034
- Tested for homogeneity and long-term stability using GC-MS and GC-FID
- Supplied with a comprehensive certificate including overall uncertainty as well as the individually determined uncertainties of all the components
Components:
α-Amylcinnamyl alcohol
trans-Anethole
4-Methoxybenzyl alcohol
Benzyl alcohol
β-Caryophyllene
Cinnamyl alcohol
Citronellol
Ebanol total
Eugenol
trans,trans-Farnesol
Geraniol
Isoeugenol (E+Z)
(R)-(+)-Limonene
Linalool
Menthol
(+)-α-Pinene
(−)-β-Pinene
β-Santalol
Sclareol
α-Terpinene
Terpineol total
tert-Butyl methyl ether
β,β,3-Trimethylbenzenepropanol
Specifications
- M-Clarity Qualitätsstufe:MQ300
- Lagertemperatur:−20°C
- Typ:Multi-component solution
Specifications
Frequently Bought Together