ContactUsLinkComponent
Hand-Operated Water Transfer Pump, 6 strokes/gallon
Water Pump, 6 strokes/gallon, 3 foot inlet tube
Catalog # MFLX70607-14
undefined
Water Pump, 6 strokes/gallon, 3 foot inlet tube
Catalog # MFLX70607-14
Specifications
  • Flow rate:
    6 strokes/gal.
  • Description:
    Water Pump, 6 strokes/gallon, 3 foot inlet tube
  • Wetted materials:
    PVC body, hose and shaft
  • Tube length, inlet:
    36"
  • Warranty:
    1 Month
  • Pk:
    1
Specifications
About this item
Ideal for removing water in municipal and irrigation markets in addition to facilities maintenance.
  • Recommended for aqueous solutions
  • Withstand temperatures up to 120F (49C)
  • Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping water and aqueous solutions. Constructed with a PVC body, hose and shaft, this pump offers 6 strokes/gallon.
  • Flow rate:
    6 strokes/gal.
  • Description:
    Water Pump, 6 strokes/gallon, 3 foot inlet tube
  • Environmentally Preferable:
  • Wetted materials:
    PVC body, hose and shaft
  • Tube length, inlet:
    36"
  • Warranty:
    1 Month
  • Pk:
    1