Orders LinkContactUsLinkComponent
Bulkhead Fitting Connectors, PTFE
Bulkhead Fitting Connectors, PTFE
Bulkhead Fitting, PTFE, ⅛" NPT(F) Portto1⅝" OD, Compression to Thread, PTFE, Straight, ⅛"
Catalog # MFLX34031-11
Bulkhead Fitting Connectors, PTFE
Catalog # MFLX34031-11
Bulkhead Fitting Connectors, PTFE
Bulkhead Fitting Connectors, PTFE
Bulkhead Fitting, PTFE, ⅛" NPT(F) Portto1⅝" OD, Compression to Thread, PTFE, Straight, ⅛"
Price
254,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Suitable for use with flat-wall or most cylindrical, smooth-wall tanks.
  • Includes FEP encapsulated silicone O-ring
  • Body can be made longer to accommodate thicker tank walls
Constructed of durable PTFE, these Bulkhead Fitting Connectors are ideal for a wide range of applications.
Specifications
  • Connection Type:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 2):
    ⅛"
  • Description:
    Bulkhead Fitting, PTFE, ⅛" NPT(F) Portto1⅝" OD
  • Fitting material:
    PTFE
  • Bulkhead outer diameter:
    1,625
  • Cat. No.:
    MFLX34031-11
  • Warranty:
    2 years
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)