ASCO® Subminiature Solenoid Valves, Stainless Steel
Constructed of stainless steel (SS) for enhanced durability.
- Two-way normal closed operation
- Install in any position
- Available with manual operator
These ASCO® subminiature solenoid valves feature a compact design that can be installed in any position. Choose from 120 VAC or 12 VDC options.
- Environmentally Preferable:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.