About this item
Complete, self-contained system for metering precise amounts of fluid.
- Maximum flow up to 480 ml/min│Durable polyethylene tank with extra heavy wall thickness for enhanced strength│Available in 15 and 30 gallon capacities
- Unit fits easily in any out-of-the-way corner
- Top-mounted, multi-position tank opening — leave open, closed or vented
Metering, injector features heavy-duty, double ball valve pump head — enhanced priming capabilities make it exceptional for purging air and gas.
- Environmentally Preferable:
Some Products May Appear Restricted