ContactUsLinkComponent
VWR®, Spill Kit, Base Neutralising
VWR®, Spill Kit, Base Neutralising
Catalog # 121-0197
Supplier:  VWR Collection
undefined
VWR®, Spill Kit, Base Neutralising
Catalog # 121-0197
Supplier:  VWR Collection
Specifications
  • Weight:
    2,5 kg
  • Description:
    Spill kit, base neutralising
  • Absorbency:
    17 L
  • Pk:
    1 KIT
Specifications
About this item
This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tub, providing 17 litres of absorption capacity.
  • Use to neutralise highly basic substances before clean-up
  • Lightweight and easy to move for a fast response
  • Highly absorbing pads to wipe down equipment and remove spills
  • SOC’s to prevent spills from spreading on the work floor
  • Neutralising polymer to neutralise bases
  • Gloves and glasses to protect employees
  • Waste disposal bag to contain all used equipment
Position spill kits around your facility so that you are prepared should a spill occur. These kits combine pads, accessories and PPE so that you are prepared in an emergency.
This spill kit contains chemical neutralising polymers to neutralise highly basic substances before clean-up, with a colour change indicator that reflects neutralisation. Also included in each spill kit are practical, visual instructions for optimal product use.
Delivery: Each kit contains 2 waste bags, 1 pair of gloves, 2 VWR® universal socs (121-0144), 10 VWR® universal pads (121-0138), 1 safety goggle, 1 overall, 1 base neutraliser, 1 scoop and scraper. With practical, visual instructions for optimal product use.