Media IDMEM W/GLN+HPS power, Corning®
Catalog # 50-016-PB
Supplier: Corning
CAS Number:
Specifications
- Media Format:Powder
- Pk:1
- Description:Iscove’s Modified DMEM: with L-glutamine and 25 mM HEPES, without [beta]-thioglycerol, [beta]-mercaptoethanol, and sodium bicarbonate