Poloxamer, Corning®
: Corning
Poloxamer 188 is a surfactant used in cell culture to protect cells in suspension against possible damage during transfer, freezing and thawing, and stirring.
It is a non-ionic detergent with an average molecular weight of 8400Da consisting of a central polypropylene flanked by two polyethylene molecules.
- Storage temperature:Refrigerate