Pre-coated PAMPA plate system, Corning® BioCoat™
Catalog # BDAA353015
Supplier: Corning
Specifications
- Membrane:PVDF
- Description:96-Well pre-coated filter plate
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:ISO 14001 - Bedford
- Pk:5
Specifications
About this item
A 96-well, pre-coated filter plate, individually packaged with a 96-well microassay receiver plate.
- 96-well insert system with a filter plate pre-coated with structured layers of phospholipids
- Configuration complies with ANSI standards
- Polystyrene plate with 0,4 µm Polyvinylidene fluoride membrane