VWR®, Spill Kit Chemical, 16 L
121-0194
- Weight:1,8 kg
- Colour:Yellow
- Description:Spill kit for chemicals
- W×D×H:500×400×110 mm
- Absorbency:16 L
- Pk:1 KIT
This portable, economy spill kit comes in a high visibility, water-resistant, yellow PVC bag with handles. Designed for small spills of chemicals and hazardous liquids. Ideal for small storage areas, thanks to its compact size.
- Fast spill response
- Yellow colour for high visibility
- Water-resistant, lightweight bag keeps sorbents dry
- Can fit behind or under a seat in a vehicle
All you need for an emergency spill response in a compact and high vis carry bag. Ideal for workers on the go, like field workers, or for carriers and truck drivers who occasionally transport small quantities of liquids.
: Every kit is supplied with 10 VWR® pads (41×51 cm), 2 VWR® socks (Ø 7,6×122 cm), 1 pair of gloves, 1 disposal bag and a 10 step plan also 'what to do when a spill occurs'.
: Packed in a durable, yellow PVC bag with handles.