Eppendorf MiniSpin and MiniSpin Plus Centrifuges
Supplier: Avantor Fluid Handling
Certificates
About this item
Small enough for a centrifuge at every workstation!
- Extremely compact footprint — measures 9"W x 4¾"H x 9½"D!
- Aluminum rotor accommodates twelve 1.5 or 2.0 ml tubes (maximum diameter of 11 mm)
- Rotor can be autoclaved at 121 °C for 20 minutes
- Minimal sample heat transfer — maximum of 12 °C after 20 minutes at max speed
- Eliminate costly repairs with the low maintenance drive
- Soft-touch lid closure for ergonomic lid locking — and releases automatically at the end of a run
- Environmentally Preferable:
Some Products May Appear Restricted