ContactUsLinkComponent
Flux Pumps Sanitary Drum Pump Air Motors
Flux Pumps Sanitary Drum Pump Air Motors
Certificates
undefined
Flux Pumps Sanitary Drum Pump Air Motors
Specifications
  • Air consumption:
  • 28,9 m³/h (17 ft.³/min)
  • Max. viscosity:
  • 1200 cP
  • 1200 cP
  • Motor hp:
  • 1/2
  • 1/2
  • Warranty:
  • 6 Months
  • 6 Months
Specifications