ContactUsLinkComponent
Agarose LE, molecular biology grade, Axygen®
Agarose LE, molecular biology grade, Axygen®
Supplier:  Corning
   Sustainable Options Available
Certificates
Agarose LE, molecular biology grade, Axygen®
Supplier:  Corning
   Sustainable Options Available
Specifications
  • Catalog No:
  • AXYGAGR-LE-1001
  • AXYGAGR-LE-5001
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Gelling temperature:
  • 36 °C (±1,5 °C) [1,5%]
  • 36 °C (±1,5 °C) [1,5%]
  • Melting point:
  • 88°C (±1,5°C) [1,5%]
  • 88°C (±1,5°C) [1,5%]
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications