ESD safety ankle boots, FINJA XXTL, Mid, S3
Supplier: ELTEN
Certificates
About this item
This shoe is ideally tailored to the ergonomics of female feet. Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
- Cowhide leather
- Fabric lining for natural ventilation
- Full, padded bellows tongue
- Full-length inlay sole ESD PRO Lady
- Non-metallic, penetration-resistant midsole
- PU/PU sole WELLMAXX Trainers Lady
- Abrasion-proof toe protection
- Steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.
Specifications
- Catalog No:
- ELTN746181/35
- ELTN746181/37
- ELTN746181/36
- ELTN746181/39
- ELTN746181/38
- ELTN746181/41
- ELTN746181/40
- ELTN746181/42
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001