Gel documentation systems, E-BOX CX5 TS Edge
The E-Box CX5 TS Edge is a modern and fully automated stand alone gel documentation instrument with innovative and outstanding technical features. By the push of only one button high-resolution images of DNA, RNA or protein gels are created within a few seconds ('One-Button-Imaging').
- Easy to use
- Stand alone gel documentation instrument with 12.4" multi-function display
- Scientific grade Geldoc camera with 5 MP resolution and 6x zoom lens
- Robust and lightproof cabinet
Thanks to its Scientific Grade 5 MP Geldoc camera and fast lens with 6× zoom the E-Box™ CX5 TS achieves an unprecedented sensitivity with a filter size of up to 210×260 mm. The large 12,4" display is made of shatterproof glass. The robust and lightproof cabinet is made totally of high quality metal.
Ordering information: Supplied without UV table for customized configuration with UV pads.
Delivery information: For full details about options available with this system, please contact your local Avantor sales office.
- Description:E-BOX CX5 TS Edge without UV table/pads
Complementary Products
- ModelWavelengthFilter size