ContactUsLinkComponent
Dumont #2, laminectomy forceps
Dumont #2, laminectomy forceps
Catalog # FSCI11223-20
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #2, laminectomy forceps
Catalog # FSCI11223-20
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    #2
  • Length:
    120 mm
  • Material:
    Inox
  • Tip style:
    Standard
  • Pk:
    1
  • Tip size:
    0,4×0,02 mm
  • Tip shape:
    Straight