ContactUsLinkComponent
Dumont #5, ceramic coated forceps
Dumont #5, ceramic coated forceps
Catalog # FSCI11252-50
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #5, ceramic coated forceps
Catalog # FSCI11252-50
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    #5
  • Length:
    110 mm
  • Material:
    Inox, ceramic coated
  • Tip style:
    Biology
  • Pk:
    1
  • Tip size:
    0,05×0,01 mm
  • Tip shape:
    Straight