ContactUsLinkComponent
Dumont #5AC anti-capillary forceps
Dumont #5AC anti-capillary forceps
Catalog # FSCI11258-20
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #5AC anti-capillary forceps
Catalog # FSCI11258-20
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    #5 anti-capillary
  • Length:
    115 mm
  • Material:
    Inox
  • Tip style:
    Biology
  • Pk:
    1
  • Tip size:
    0,07×0,02 mm
  • Tip shape:
    Straight