Order Entry
Germany
ContactUsLinkComponent
London forceps
London forceps
Catalog # FSCI11080-02
Supplier:  FINE SCIENCE TOOLS
undefined
London forceps
Catalog # FSCI11080-02
Supplier:  FINE SCIENCE TOOLS

Specifications

  • Length:
    160 mm
  • Material:
    Stainless steel
  • Tip style:
    Serrated
  • Pk:
    1
  • Tip size:
    1×0,9 mm
  • Tip shape:
    Angled (45°)

Specifications

About this item