Order Entry
Germany
Orders LinkContactUsLinkComponent
Student fine forceps, curved

Student fine forceps, curved

Fine, Stainless steel, 100 mm, Standard, Curved, 0,2×0,2 mm
Catalog # FSCI91117-10
Marketsource
Supplier:   FINE SCIENCE TOOLS
Student fine forceps, curved
Catalog # FSCI91117-10
Supplier:  FINE SCIENCE TOOLS
Marketsource
Student fine forceps, curved
Student fine forceps, curved

Fine, Stainless steel, 100 mm, Standard, Curved, 0,2×0,2 mm

Price
59,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Fine
  • Length:
    100 mm
  • Material:
    Stainless steel
  • Tip style:
    Standard
  • Tip size:
    0,2×0,2 mm
  • Tip shape:
    Curved
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.