ContactUsLinkComponent
Silver neuro clips
Silver neuro clips
Catalog # FSCI18040-00
Supplier:  FINE SCIENCE TOOLS
undefined
Silver neuro clips
Catalog # FSCI18040-00
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Description:
    Silver Neuro Clips
  • Size:
    5 mm
  • Pk:
    100
Specifications
About this item
These single-use silver neuro clips are meant for permanent vessel occlusion.
  • Clamp Press: Variable
  • Alloy / Material: Silver
  • Length: 5 mm
  • Not serrated
Caution: All surgical instruments and devices from the Fine Science Tools product range are intended exclusively for use in experimental research laboratories and facilities or in veterinary medicine.