ContactUsLinkComponent
Surgical scissors, serrated
Surgical scissors, serrated
Catalog # FSCI14007-14
Supplier:  FINE SCIENCE TOOLS
undefined
Surgical scissors, serrated
Catalog # FSCI14007-14
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    Sharp/blunt, straight tips
  • Length:
    140 mm
  • Pk:
    1
  • Serrated:
    +
  • Blade length:
    40 mm
Specifications
About this item
These scissors are made of stainless steel.