Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Scissors
Mini Lister scissors, Tungsten Carbide
Mini Lister scissors, Tungsten Carbide
Mini Lister scissors, Tungsten Carbide
Catalog # FSCI14531-09
Supplier:
FINE SCIENCE TOOLS
Expand image
Mini Lister scissors, Tungsten Carbide
Catalog # FSCI14531-09
Supplier:
FINE SCIENCE TOOLS
Specifications
Type:
Blunt/blunt, angled to side tips, smooth, tungsten carbide
Length:
90 mm
Pk:
1
Specifications
About this item
Product Details
Product Details