ContactUsLinkComponent
Surgical scissors, large loops
Surgical scissors, large loops
Catalog # FSCI14101-14
Supplier:  FINE SCIENCE TOOLS
undefined
Surgical scissors, large loops
Catalog # FSCI14101-14
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    Sharp/blunt, straight tips, large loops
  • Length:
    145 mm
  • Pk:
    1
  • Blade length:
    45 mm
Specifications
About this item
These scissors are made of stainless steel.