Orders LinkContactUsLinkComponent
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt
Marketsource
Supplier:   FINE SCIENCE TOOLS
Surgical scissors, sharp-blunt
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Surgical scissors, sharp-blunt
Surgical scissors, sharp-blunt
10 Options Available Below
Product Details & Documents
These scissors are made of stainless steel.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Availability
    Price
  • Sharp/blunt, straight tips
    120 mm
    41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 60,60 €
    60,60 €
    1 * 1 ST
       60,60 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, curved tips
    120 mm
    41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,00 €
    74,00 €
    1 * 1 ST
       74,00 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, straight tips
    130 mm
    42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 61,50 €
    61,50 €
    1 * 1 ST
       61,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, curved tips
    130 mm
    42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 72,10 €
    72,10 €
    1 * 1 ST
       72,10 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, curved tips
    145 mm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,90 €
    75,90 €
    1 * 1 ST
       75,90 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, straight tips, left-handed
    145 mm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 258,00 €
    258,00 €
    1 * 1 ST
       258,00 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, straight tips
    145 mm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,40 €
    64,40 €
    1 * 1 ST
       64,40 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, curved tips
    165 mm
    53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,70 €
    80,70 €
    1 * 1 ST
       80,70 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, curved tips
    185 mm
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,50 €
    86,50 €
    1 * 1 ST
       86,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp/blunt, straight tips
    185 mm
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 86,50 €
    86,50 €
    1 * 1 ST
       86,50 €
    Specifications:
    Marketsource
    Marketsource