Instrument trays, stainless steel
Supplier: BURKLE
Certificates
About this item
Instruments, labware or samplers can be placed and transported on the sturdy stainless steel tray. The smooth surface is easy to clean.
- Stainless steel V2A
- Prepare, place and carry instruments and samples
- Hygienic and robust
The rectangular instrument tray is suitable for hygienic placing in the fields of chemistry, pharmaceuticals, cosmetics, animal feed and food and beverage.